missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptylamine, 99%, Thermo Scientific Chemicals

Product Code. 10234040
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5452-35-7
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.2
MDL Number MFCD00004153
InChI Key VXVVUHQULXCUPF-UHFFFAOYSA-N
Synonym cycloheptylamine, aminocycloheptane, cycloheptyl amine, cycloheptanamine, n-cycloheptylamine, 1-cycloheptylamine, cycloheptanyl amine, sz1, acmc-209lh8
PubChem CID 2899
IUPAC Name cycloheptanamine
SMILES C1CCCC(CC1)N

Specifications

Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 54.0°C (11.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C7H13NH2
Refractive Index 1.4710 to 1.4730
Quantity 25 g
Beilstein 12, 8
Specific Gravity 0.89
Solubility Information Solubility in water: soluble.
Formula Weight 113.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cycloheptylamine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cycloheptylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'