missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, 99.8%, for HPLC

Product Code. 10053390 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10053390

Brand: Thermo Scientific 279590025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-82-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifica

Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.004 max. at 240nm, 0.004 max. at 250nm, 0.018 max. at 230nm, 0.097 max. at 220nm, 0.201 max. at 210nm, 0.301 max. at 200nm
Color White
Density 0.7700g/mL
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Crystalline Solid
Assay Percent Range 99.8%
Residue after Evaporation 0.0002 % max.
Linear Formula C6H12
Vapor Pressure Vapor Pressure: 104mbar at 20°C
Beilstein 05, 20
Water 0.01% Max.
Merck Index 15, 2716
Formula Weight 84.15
Percent Purity ≥99.8% (GC)
Grade HPLC
Chemical Name or Material Cyclohexane
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons compliance-icons compliance-icons
Identificativo prodotto
  • Cyclohexane
Avvertenza
  • Danger
Categoria di pericolo
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Indicazioni di pericolo
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato