missing translation for 'onlineSavingsMsg'
Learn More
Cyclohexane, 99+%, pure, Thermo Scientific Chemicals Product Code.: 10778893

Cyclohexane, 99+%, pure, Thermo Scientific Chemicals

Product Code. 10778893
10 L, Plastic Drum
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Glass Bottle
Plastic Drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10778893

Brand: Thermo Scientific Acros 111110100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Density 0.7700g/mL
Boiling Point 81.0°C
Quantity 10 L
Flash Point −18°C
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.4250 to 1.4270
Assay Percent Range 99+%
Residue after Evaporation 0.005% max.
Specific Gravity 0.77
Linear Formula C6H12
Vapor Pressure Vapor Pressure: 104mbar at 20°C
Viscosity 0.94 mPa.s (20°C)
Beilstein 05, 20
Water 0.05% max. (K.F.)
Merck Index 15, 2716
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
Formula Weight 84.15
Percent Purity ≥99%
Grade Pure
Chemical Name or Material Cyclohexane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2495548 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number111110250,111110100,111110010,111110025,11111044
Lot Number2412835 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number111110250,111110100,111110010,11111044,111110025
Lot Number2523811 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number111110250,111110050,111110100,111110010,111110025
Lot Number2523811 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number111110250,111110100,111110010,111110025,11111044
Lot Number2522628 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number111110250,111110100,111110010,111110025,11111044
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexane, 99+%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.