missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, BAKER, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
Description
A grade of chemicals for routine research usually controlled by physical criteria and often by assay (by GC).
Specifications
Specifications
| Melting Point | 6.4°C |
| Color | Colorless |
| Density | 0.78g/cm3 at 20°C |
| Boiling Point | 81°C |
| Quantity | 2.5 L |
| Explosion Limit | 1.2 - 8.3 Vol % |
| Flash Point | −18°C |
| Ignition Point | 260°C |
| Odor | Sweetish |
| Packaging | Glass |
| Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P331-Do NOT induce vomiting.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction