missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riedel-de Haën Cyclohexane CHROMASOLV™
Click to view available options
Quantity:
7 L
Unit Size:
7 L
Description
- Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy.
Specifications
Specifications
| pH | Not applicable |
| Melting Point | 6.4°C |
| CAS | 110-82-7 |
| Color | Colorless |
| Density | 0.78 g/cm3 (20°C) |
| Boiling Point | 81°C (1.013 hPa) |
| Quantity | 7 L |
| Flash Point | 8.3 %(V) |
| Packaging | Returnable Stainless Steel Can |
| Physical Form | Liquid |
| Show More |
Safety and Handling
Product Identifier
- Cyclohexane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction