missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, Extra Pure, SLR, Fisher Chemical™
Sfoglia Tutto Fisher Chemical Prodotti
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
HDPE Plastic Bottle
Metal can
Plastic Coated Glass bottle
Plastic container
Dimensione della confezione:
2.5 L
25 L
5 L
500mL
Specifica
Specifica
| Packaging | Plastic Coated Glass bottle |
| CAS | 110-82-7 |
| Vapor Pressure | 104mbar at 20°C |
| Viscosity | 0.94 mPaS at 20°C |
| Vapor Density | 2.9 |
| Physical Form | Liquid |
| Molecular Formula | C6H12 |
| MDL Number | 3814 |
| Color | Colorless |
| Melting Point | 6.5°C |
| Vedi altri risultati |
Sicurezza e movimentazione
Identificativo prodotto
- Cyclohexane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto