missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical C/8920/PB08

24.50 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.



 View more versions of this product

Product Code. 11458143

  • £35.85 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

110-82-7
500 mL
C6H12
84.15g/mol
0.94 mPaS at 20°C
6.5°C
Liquid
Plastic Coated Glass bottle
Colorless
3814
104mbar at 20°C
2.9
81°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cyclohexane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers