missing translation for 'onlineSavingsMsg'
Learn More
Cyclohexane (HPLC), Fisher Chemical™ Product Code.: 10766091

Cyclohexane (HPLC), Fisher Chemical™

Product Code. 10766091
2.5 L, Amber Glass Bottle
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10766091

Brand: Fisher Chemical C/8936/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Color Colorless
Boiling Point 81°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 104mbar at 20°C
Viscosity 0.94 mPaS at 20°C
Formula Weight 84.15g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2526771 Certificate TypeCertificate of Analysis Date13/02/2025 Catalog NumberC/8936/25RSS,C/8936/10RSS,C/8936/27SS,C/8936/PB17,C/8936/30RSS,C/8936/15,C/8936/17
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexane (HPLC), Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.