missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10173380
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-89-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001461
InChI Key NZNMSOFKMUBTKW-UHFFFAOYSA-N
Synonym hexahydrobenzoic acid, carboxycyclohexane, cyclohexanoic acid, cyclohexylcarboxylic acid, cyclohexylmethanoic acid, cyclohexylformic acid, benzoic acid, hexahydro, cyclohexancarbonsaeure, cyclohexane-1-carboxylate, cyclohexanecarboxylicacid
PubChem CID 7413
ChEBI CHEBI:36096
IUPAC Name cyclohexanecarboxylic acid
SMILES C1CCC(CC1)C(=O)O

Specifications

Melting Point 30.0°C to 32.0°C
Color White
Density 1.0300g/mL
Boiling Point 232.0°C to 233.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H11CO2H
Refractive Index 1.4600 to 1.4620
Quantity 25 g
Beilstein 09, 7
Merck Index 15, 2717
Specific Gravity 1.03
Solubility Information Solubility in water: 0.201g/100mL (15 c). Other solubilities: soluble in alcohol and ether
Formula Weight 128.17
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material Cyclohexanecarboxylic acid, 98+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexanecarboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.