missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone 2,4-dinitrophenylhydrazone, 99%, Thermo Scientific Chemicals

Product Code. 11404873
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cyclohexanone 2,4-dinitrophenylhydrazone is a useful standard for elemental analysis. Also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanone 2,4-dinitrophenylhydrazone is a useful standard for elemental analysis. Also used as a pharmaceutical intermediate.

Solubility
Soluble in water (partly).

Notes
Store at 2-8°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1589-62-4
Molecular Formula C12H14N4O4
Molecular Weight (g/mol) 278.268
MDL Number MFCD00001658
InChI Key QLWXZRVOHCYKKK-UHFFFAOYSA-N
Synonym cyclohexanone 2,4-dinitrophenylhydrazone, 1-cyclohexylidene-2-2,4-dinitrophenyl hydrazine, cyclohexanone-2,4-dinitrophenylhydrazone, cyclohexanone, 2,4-dinitrophenyl hydrazone, cyclohexanone 2,4-dinitrophenyl hydrazone, n-cyclohexylideneamino-2,4-dinitroaniline, acmc-20alct, maybridge1_001626
PubChem CID 74117
IUPAC Name N-(cyclohexylideneamino)-2,4-dinitroaniline
SMILES C1CCC(=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-])CC1

Specifications

Melting Point 158°C to 160°C
Quantity 5 g
Solubility Information Soluble in water (partly).
Formula Weight 278.27
Percent Purity 99%
Chemical Name or Material Cyclohexanone 2,4-dinitrophenylhydrazone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'