missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanone 2,4-dinitrophenylhydrazone, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Cyclohexanone 2,4-dinitrophenylhydrazone is a useful standard for elemental analysis. Also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclohexanone 2,4-dinitrophenylhydrazone is a useful standard for elemental analysis. Also used as a pharmaceutical intermediate.
Solubility
Soluble in water (partly).
Notes
Store at 2-8°C. Incompatible with oxidizing agents.
Cyclohexanone 2,4-dinitrophenylhydrazone is a useful standard for elemental analysis. Also used as a pharmaceutical intermediate.
Solubility
Soluble in water (partly).
Notes
Store at 2-8°C. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 158°C to 160°C |
Quantity | 25 g |
Solubility Information | Soluble in water (partly). |
Formula Weight | 278.27 |
Percent Purity | 99% |
Chemical Name or Material | Cyclohexanone 2,4-dinitrophenylhydrazone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'