missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, 99+%, Thermo Scientific Chemicals

£21.91 - £131.66

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
PubChem CID 7967
ChEBI CHEBI:17854
IUPAC Name cyclohexanone
SMILES C1CCC(=O)CC1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15245998
View Documents
Thermo Scientific Acros
406092500
250 mL
£21.91
250mL
Please to purchase this item. Need a web account? Register with us today!
15204187
View Documents
Thermo Scientific Acros
406090010
1 L
£58.49
1 L
Please to purchase this item. Need a web account? Register with us today!
15235998
View Documents
Thermo Scientific Acros
406090025
2.5 L
£131.66
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-94-1
98.145
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
CHEBI:17854
C1CCC(=O)CC1
C6H10O
JHIVVAPYMSGYDF-UHFFFAOYSA-N
7967
cyclohexanone

Specifications

-47.0°C
0.9470g/mL
43°C
0.947
C6H10(=O)
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
JHIVVAPYMSGYDF-UHFFFAOYSA-N
cyclohexanone
7967
98.14
ACS Reagent
108-94-1
155.0°C
1.4450 to 1.4560
C6H10O
0.05% max.
Solubility in water: 150g/L (10°C). Other solubilities: soluble in all common organic solvents
C1CCC(=O)CC1
98.145
CHEBI:17854
≥99%
Cyclohexanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement Flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye damage. Causes skin irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

Product Identifier
  • Cyclohexanone

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.