missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone Oxime 97%, Thermo Scientific Chemicals

Catalog Number 10734411
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
100-64-1
C6H11NO
113.16
MFCD00001660
VEZUQRBDRNJBJY-UHFFFAOYSA-N
cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
7517
N-cyclohexylidenehydroxylamine
ON=C1CCCCC1
This item is not returnable. View return policy
100-64-1
C6H11NO
113.16
MFCD00001660
VEZUQRBDRNJBJY-UHFFFAOYSA-N
cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
7517
N-cyclohexylidenehydroxylamine
ON=C1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 88°C to 92°C
Color Beige-Brown to White
Quantity 500 g
Infrared Spectrum Authentic
Packaging Plastic Bottle
Physical Form Crystalline Powder, Crystals, or Chunks
Assay Percent Range 97%
Linear Formula C6H10(=NOH)
Beilstein 07, 10
Merck Index 10, 2720
Formula Weight 113.16
Percent Purity ≥96.0%
Chemical Name or Material Cyclohexanone oxime, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'