Learn More
Cyclohexene oxide, 98%, Thermo Scientific Chemicals
£32.16 - £98.29
Chemical Identifiers
CAS | 286-20-4 |
---|---|
Molecular Formula | C6H10O |
Molecular Weight (g/mol) | 98.14 |
MDL Number | MFCD00005162 |
InChI Key | ZWAJLVLEBYIOTI-UHFFFAOYSA-N |
Synonym | cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxide |
PubChem CID | 9246 |
IUPAC Name | 7-oxabicyclo[4.1.0]heptane |
SMILES | C1CCC2C(C1)O2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10510371
|
Thermo Scientific Acros
111211000 |
100 mL | Glass bottle |
£32.16
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10214330
|
Thermo Scientific Acros
111215000 |
500 mL | Glass bottle |
£98.29
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
286-20-4 | |
98.14 | |
ZWAJLVLEBYIOTI-UHFFFAOYSA-N | |
9246 | |
C1CCC2C(C1)O2 |
C6H10O | |
MFCD00005162 | |
cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxide | |
7-oxabicyclo[4.1.0]heptane |
Specifications
286-20-4 | |
Colorless to Yellow | |
129.0°C to 130.0°C | |
Authentic | |
C6H10O | |
MFCD00005162 | |
0.97 | |
Solubility in water: insoluble in water. Other solubilities: soluble in alcohol, ether and acetone, soluble in most other organic solvents | |
C1CCC2C(C1)O2 | |
98.14 | |
1.75 mPa.s (25°C) | |
98% | |
Cyclohexene oxide, 98% |
-40.0°C | |
0.9700g/mL | |
27°C | |
97.5% min. (GC) | |
1.4520 to 1.4540 | |
17, 21 | |
cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxide | |
ZWAJLVLEBYIOTI-UHFFFAOYSA-N | |
7-oxabicyclo[4.1.0]heptane | |
9246 | |
98.14 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-007-7
missing translation for 'rtecsNumber' : RN7175000
missing translation for 'tsca' : TSCA
- Cyclohexene oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.