missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Cycloheximide, 95% Product Code.: 10159861

Thermo Scientific Chemicals Cycloheximide, 95%

Product Code. 10159861
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 10159861

Brand: Thermo Scientific Chemicals 357420050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cycloheximide, CAS # 66-81-9, is a compound produced by the bacterium Streptomyces griseus that shows an antifungal antibiotic activity with the ability to inhibit protein synthesis only in eukaryotic cells.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cycloheximide is a semisynthetic compound showing an antifungal antibiotic activity.
  • It is produced by the bacterium Streptomyces griseus
  • Cycloheximide can inhibit protein synthesis in eukaryotic cells but not in prokaryotes by interacting directly with the translocase enzyme and interfering with the translocation step. Furthermore, this compound also blocks the A site of ribosomes

Application

  • Cycloheximide shows a neuroprotective, fungicide, and anticoronaviral activities
  • In biomedical research, it can induce glycogenolysis, gluconeogenesis, and ureagenesis
  • Due to significant toxic side effects such as teratogenesis, DNA damage, birth defects, and toxicity to sperm, this chemical compound is only used in in vitro research applications
  • This compound also shows inhibitory activity against ferroptosis
  • It is also used in laboratory media as a selective agent to permit the isolation of pathogenic and non-pathogenic fungi from other types of specimens
  • In laboratory research related to plant biology, it can induce ethylene production to promote the plant growth
  • In laboratory research, this compound can be applied to determine the half-life of protein in any cellular system. The treatment of in vitro cells with cycloheximide in a time-course experiment followed by western blotting using this cell lysate can show differences in protein half-life.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-81-9
Molecular Formula C15H23NO4
Molecular Weight (g/mol) 281.35
MDL Number MFCD00082346
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
Synonym cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide
PubChem CID 6197
ChEBI CHEBI:27641
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1

Specifications

Melting Point 107.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,2721
Specific Rotation Condition − 28.50 (20.00°C c=1,CHCl3)
Specific Rotation − 28.50
Solubility Information Solubility in water: 2.1 g/100mL (2 c). Other solubilities: soluble in chloroform, ether, acetone, methanol, and ethanol
Formula Weight 281.35
Percent Purity 95%
Physical Form Glassy Irregular Shaped Granules
Chemical Name or Material Cycloheximide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467922 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number357420050,357420010,357420250
Lot NumberA0464024 Certificate TypeCertificate of Analysis Date05/07/2024 Catalog Number357420050,357420010,357420250
Lot NumberA0457904 Certificate TypeCertificate of Analysis Date23/11/2023 Catalog Number357420050,357420010,357420250
Lot NumberA0430035 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number357420050,357420010,357420250
Lot NumberA0437935 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number357420050,357420010,357420250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cycloheximide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
  • H341-Suspected of causing genetic defects.
  • H411-Toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Cycloheximide, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.