missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl 2-chloroacetate, 97%, Thermo Scientific™

Product Code. 10436662
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6975-91-3
Molecular Formula C8H13ClO2
Molecular Weight (g/mol) 176.64
MDL Number MFCD00084989
InChI Key IBFHBLOKQVCABG-UHFFFAOYSA-N
Synonym cyclohexyl chloroacetate, cyclohexyl chloroacetate #, 1-chloroacetyl oxy cyclohexane, chloroacetic acid cyclohexyl ester, chloroacetic acid, cyclohexyl ester, acetic acid, 2-chloro-, cyclohexyl ester
PubChem CID 228008
IUPAC Name cyclohexyl 2-chloroacetate
SMILES C1CCC(CC1)OC(=O)CCl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula ClCH2CO2C6H11
Quantity 10 g
Formula Weight 176.64
Percent Purity 97%
Chemical Name or Material Cyclohexyl 2-chloroacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.