missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl chloride, 98%, Thermo Scientific Chemicals

£17.90 - £89.95

Chemical Identifiers

CAS 542-18-7
Molecular Formula C6H11Cl
Molecular Weight (g/mol) 118.61
MDL Number MFCD00003822
InChI Key UNFUYWDGSFDHCW-UHFFFAOYSA-N
Synonym cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane
PubChem CID 10952
ChEBI CHEBI:39156
IUPAC Name chlorocyclohexane
SMILES C1CCC(CC1)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10325840
View Documents
Thermo Scientific
111301000
100 g Glass bottle
£17.90
100g
Please to purchase this item. Need a web account? Register with us today!
10214140
View Documents
Thermo Scientific Acros
111302500
250 g Glass bottle
£32.46
250g
Please to purchase this item. Need a web account? Register with us today!
10152470
View Documents
Thermo Scientific Acros
111300010
1 kg Glass bottle
£89.95
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

542-18-7
118.61
UNFUYWDGSFDHCW-UHFFFAOYSA-N
10952
chlorocyclohexane
C6H11Cl
MFCD00003822
cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane
CHEBI:39156
C1CCC(CC1)Cl

Specifications

542-18-7
1.0000g/mL
28°C
97.5% min. (GC)
C6H11Cl
MFCD00003822
15, 2725
cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane
UNFUYWDGSFDHCW-UHFFFAOYSA-N
chlorocyclohexane
10952
1.73 mPa.s (20°C)
98%
Cyclohexyl chloride, 98%
-44°C
142°C
Authentic
C6H11Cl
1.4610 to 1.463
05, 21
1
Solubility in water: 0.02g/L (20°C). Other solubilities: soluble in alcohol and diethyl ether
C1CCC(CC1)Cl
118.61
CHEBI:39156
118.61
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 208-806-6

missing translation for 'rtecsNumber' : GU8685000

missing translation for 'tsca' : TSCA

Product Identifier
  • Cyclohexyl chloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.