missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl hexanoate, 99%

Product Code. 11437124
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11437124

Brand: Thermo Scientific Alfa Aesar B21547.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6243-10-3 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6243-10-3
MDL Number MFCD00046356

Specifications

Molecular Formula C12H22O2
Refractive Index 1.446
Quantity 25 g
Synonym hexanoic acid, cyclohexyl ester, cyclohexyl caproate, unii-v3b77j8gnh, v3b77j8gnh, cyclohexyl n-hexanoate, hexanoic acid cyclohexyl ester, acmc-1b676, 4-4-methoxyphenyl-1,3-thiazol-4-amine
InChI Key DTNOERNOMHQUCN-UHFFFAOYSA-N
SMILES CCCCCC(=O)OC1CCCCC1
IUPAC Name cyclohexyl hexanoate
Molecular Weight (g/mol) 198.31
PubChem CID 80388
Formula Weight 198.31
Percent Purity 99%
Chemical Name or Material Cyclohexyl hexanoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.