missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methacrylate, 97%, stabilized, ACROS Organics™

Catalog Number 10486422
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 101-43-9
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C10H16O2
Linear Formula H2C=C(CH3)CO2C6H11
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
MDL Number MFCD00014292
Quantity 5 mL
Synonym cyclohexyl methacrylate, methacrylic acid, cyclohexyl ester, 2-propenoic acid, 2-methyl-, cyclohexyl ester, unii-5l9uuv9t6q, 2-methyl-2-propenoic acid cyclohexyl ester, 5l9uuv9t6q, methacrylic acid cyclohexyl ester, ageflex chma, c-hma, cyclo hexyl methacrylate
InChI Key OIWOHHBRDFKZNC-UHFFFAOYSA-N
SMILES CC(=C)C(=O)OC1CCCCC1
IUPAC Name cyclohexyl 2-methylprop-2-enoate
Stabilizer 60 - 100ppm MEHQ
Molecular Weight (g/mol) 168.24
PubChem CID 7561
Formula Weight 168.24
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Cyclohexyl methacrylate, 97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'