missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11483547
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4998-76-9
Molecular Formula C6H14ClN
Molecular Weight (g/mol) 135.64
MDL Number MFCD00050540
InChI Key ZJUGSKJHHWASAF-UHFFFAOYSA-N
Synonym cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361
PubChem CID 2723770
IUPAC Name cyclohexanamine;hydrochloride
SMILES [Cl-].[NH3+]C1CCCCC1

Specifications

Melting Point 206°C to 209°C
Odor Odorless
Quantity 500 g
Beilstein 3613002
Sensitivity Hygroscopic
Formula Weight 135.64
Percent Purity ≥98%
Chemical Name or Material Cyclohexylamine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'