missing translation for 'onlineSavingsMsg'
Learn More
Cyclohexylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles Product Code.: 15415425

Cyclohexylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15415425
50 mL
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy

Product Code. 15415425

Brand: Thermo Scientific Alfa Aesar H58852.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7565-57-3 | C6H11BrZn | 228.438 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary intermediate.

Solubility
Reacts with water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7565-57-3
Density 0.963
Molecular Formula C6H11BrZn
MDL Number MFCD00671993
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym cyclohexylzinc bromide, cyclohexylzinc bromide solution, bromozinc 1+ ,cyclohexane, bromo cyclohexyl zinc, cyclohexylzinc bromide, 0.50 m in thf, cyclohexylzinc bromide solution, 0.5 m in thf, cyclohexylzinc bromide 0.5 m in tetrahydrofuran, cyclohexylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key PVURAUIMVICLOH-UHFFFAOYSA-M
SMILES C1CC[CH-]CC1.[Zn+]Br
IUPAC Name bromozinc(1+);cyclohexane
Molecular Weight (g/mol) 228.438
PubChem CID 4385789
Formula Weight 228.44
Chemical Name or Material Cyclohexylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.