Learn More
Specac™ Cyclone™ C10 Long Pathlength Gas Cell GKFV
Brand: Specac™
Code : Z3
Additional Details : Weight : 7.00000kg
Description
This is a heatable long pathlength gas cell with glass body, KBr windows, fixed pathlength and Viton O-rings. The cell has a volume of 2.61 litres.Specifications
Fixed pathlength and Viton O-rings | |
2.61 L |