missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Cyclooctatetraene)iron tricarbonyl, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
Description
(Cyclooctatetraene)iron tricarbonyl is used as primary and secondary intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 93°C to 95°C |
Color | Red to Brown |
Quantity | 0.5 g |
UN Number | UN3466 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 244.03 |
Percent Purity | 98% |
Physical Form | Crystalline |
Chemical Name or Material | (Cyclooctatetraene)iron tricarbonyl |
Safety and Handling
Product Identifier
- (Cyclooctatetraene)iron tricarbonyl
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'