Learn More
Cyclooctylamine, 97%, Thermo Scientific Chemicals
£25.10 - £200.00
Chemical Identifiers
CAS | 5452-37-9 |
---|---|
Molecular Formula | C8H17N |
Molecular Weight (g/mol) | 127.23 |
MDL Number | MFCD00001748 |
InChI Key | HSOHBWMXECKEKV-UHFFFAOYSA-N |
Synonym | cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc |
PubChem CID | 2903 |
IUPAC Name | cyclooctanamine |
SMILES | C1CCCC(CCC1)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10775941
|
Thermo Scientific Acros
111460050 |
5 g | Glass bottle |
£25.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10172420
|
Thermo Scientific Acros
111460250 |
25 g | Glass bottle |
£73.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10646914
|
Thermo Scientific Acros
111461000 |
100 g | Glass Bottle |
£200.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5452-37-9 | |
127.23 | |
HSOHBWMXECKEKV-UHFFFAOYSA-N | |
2903 | |
C1CCCC(CCC1)N |
C8H17N | |
MFCD00001748 | |
cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc | |
cyclooctanamine |
Specifications
5452-37-9 | |
Colorless to Yellow | |
190°C | |
Authentic | |
C8H17N | |
1.4800 to 1.482 | |
12, IV, 128 | |
cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc | |
HSOHBWMXECKEKV-UHFFFAOYSA-N | |
cyclooctanamine | |
2903 | |
127.23 | |
Liquid |
-48°C | |
0.9200g/mL | |
62°C | |
96% min. (GC) | |
C8H15NH2 | |
MFCD00001748 | |
0.92 | |
Solubility in water: 10.2g/L (20°C) | |
C1CCCC(CCC1)N | |
127.23 | |
5 mPa.s (20°C) | |
97% | |
Cyclooctylamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 226-694-7
- Cyclooctylamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only