missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienyltitanium trichloride, Thermo Scientific Chemicals

£41.70 - £157.00

Chemical Identifiers

CAS 1270-98-0
Molecular Formula C5H5Cl3Ti
Molecular Weight (g/mol) 219.31
MDL Number MFCD00003724
InChI Key QOXHZZQZTIGPEV-UHFFFAOYSA-K
Synonym Trichlorocyclopentadienyltitanium
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11324899
View Documents
Thermo Scientific Alfa Aesar
077134.03
1 g
£41.70
1g
Please to purchase this item. Need a web account? Register with us today!
11314899
View Documents
Thermo Scientific Alfa Aesar
077134.06
5 g
£157.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Precursor for cyclopentadienyltitanium(IV) complexes, and used as catalysts for alkene polymerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1270-98-0
219.31
QOXHZZQZTIGPEV-UHFFFAOYSA-K
C5H5Cl3Ti
MFCD00003724
Trichlorocyclopentadienyltitanium

Specifications

210°C (decomposition)
Yellow to Orange
C5H5Cl3Ti
UN3261
QOXHZZQZTIGPEV-UHFFFAOYSA-K
219.35
Cyclopentadienyltitanium trichloride
1270-98-0
Crystalline
MFCD00003724
Trichlorocyclopentadienyltitanium
219.31
Moisture Sensitive
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'rtecsNumber' : XR1927300

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only