missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentane, Reagent Grade, 98%, Honeywell

Reagent Grade, 98%

£28.06 - £274.00

Chemical Identifiers

CAS 287-92-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00001356
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
PubChem CID 9253
ChEBI CHEBI:23492
IUPAC Name cyclopentane
SMILES C1CCCC1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15695600
View Documents
Honeywell Research Chemicals
459747-100ML
100 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15625610
View Documents
Honeywell Research Chemicals
459747-500ML
500 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15615610
View Documents
Honeywell Research Chemicals
459747-2L
2 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

287-92-3
70.135
RGSFGYAAUTVSQA-UHFFFAOYSA-N
9253
cyclopentane
C5H10
MFCD00001356
pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
CHEBI:23492
C1CCCC1

Specifications

287-92-3
Colorless
50°C
682°F
C5H10
UN1146
pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
C1CCCC1
70.135
CHEBI:23492
5.3psi (21°C), 9.9psi (38°C)
Cyclopentane
-94°C
0.751g/mL at 25°C
−20°C (−4°F)
Solvent like
MFCD00001356
1900195
RGSFGYAAUTVSQA-UHFFFAOYSA-N
cyclopentane
9253
70.13g/mol
98%
Videos
Safety and Handling

Safety and Handling

P210-P273

missing translation for 'dotInformation' : class 3 - PG 2 - Cyclopentane

missing translation for 'rtecsNumber' : 206-016-6

SDS
Documents

Documents

Product Certifications