missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarbonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11458116
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cyclopentanecarbonyl chloride is used in biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanecarbonyl chloride is used in biochemical for proteomics research.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4524-93-0
Molecular Formula C6H9ClO
Molecular Weight (g/mol) 132.587
MDL Number MFCD00001370
InChI Key WEPUZBYKXNKSDH-UHFFFAOYSA-N
Synonym cyclopentanecarbonylchloride, cyclopentane carboxyl chloride, cyclopentanoyl chloride, pubchem13237, acmc-1csoo, cyclopentylformyl chloride, cyclopetanecarbonylchloride, cyclopentancarbonyl chloride, cyclopentylcarbonyl chloride, chlorocarbonyl cyclopentane
PubChem CID 78284
IUPAC Name cyclopentanecarbonyl chloride
SMILES C1CCC(C1)C(=O)Cl

Specifications

Density 1.091
Boiling Point 160°C to 162°C
Flash Point 60°C (140°F)
Odor Acrid
Refractive Index 1.464
Quantity 25 g
UN Number UN2920
Beilstein 1237147
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 132.59
Percent Purity 98%
Chemical Name or Material Cyclopentanecarbonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'