Learn More
Cyclopentanecarboxylic acid, 99%, Thermo Scientific Chemicals
£31.30 - £472.00
Chemical Identifiers
CAS | 3400-45-1 |
---|---|
Molecular Formula | C6H10O2 |
Molecular Weight (g/mol) | 114.144 |
MDL Number | MFCD00001371 |
InChI Key | JBDSSBMEKXHSJF-UHFFFAOYSA-N |
Synonym | cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g |
PubChem CID | 18840 |
IUPAC Name | cyclopentanecarboxylic acid |
SMILES | C1CCC(C1)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11419523
|
Thermo Scientific Alfa Aesar
A12375.09 |
10 g |
£31.30
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429523
|
Thermo Scientific Alfa Aesar
A12375.18 |
50 g |
£107.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439523
|
Thermo Scientific Alfa Aesar
A12375.30 |
250 g |
£472.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3400-45-1 | |
114.144 | |
JBDSSBMEKXHSJF-UHFFFAOYSA-N | |
18840 | |
C1CCC(C1)C(=O)O |
C6H10O2 | |
MFCD00001371 | |
cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g | |
cyclopentanecarboxylic acid |
Specifications
3400-45-1 | |
1.052 | |
93°C (199°F) | |
1.454 | |
2039729 | |
JBDSSBMEKXHSJF-UHFFFAOYSA-N | |
cyclopentanecarboxylic acid | |
18840 | |
99% |
-7°C | |
215°C to 216°C | |
C6H10O2 | |
MFCD00001371 | |
cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g | |
C1CCC(C1)C(=O)O | |
114.144 | |
114.14 | |
Cyclopentanecarboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 222-269-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only