missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone-2,2,5,5-d{4}, 95%, Thermo Scientific Chemicals

Product Code. 15457977
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3997-89-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 88.142
MDL Number MFCD00190415
InChI Key BGTOWKSIORTVQH-KHORGVISSA-N
Synonym cyclopentanone-2,2,5,5-d4, cyclopentanone d4, 2,2,5,5-∼2∼h_4_ cyclopentanone, cyclopentanone-2,2,5,5-d4, 98 atom % d, 2,2,5,5-2 h? cyclopentan-1-one
PubChem CID 16213326
IUPAC Name 2,2,5,5-tetradeuteriocyclopentan-1-one
SMILES C1CCC(=O)C1

Specifications

Quantity 1 g
UN Number UN2245
Formula Weight 88.14
Percent Purity 95%
Chemical Name or Material Cyclopentanone-2,2,5,5-d4

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'