missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone, 99+%, pure, Thermo Scientific Chemicals

Product Code. 10736521
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-92-3
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001409
InChI Key BGTOWKSIORTVQH-UHFFFAOYSA-N
Synonym adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy
PubChem CID 8452
ChEBI CHEBI:16486
IUPAC Name cyclopentanone
SMILES O=C1CCCC1

Specifications

Melting Point -51.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 130.0°C to 131.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C5H8(=O)
Refractive Index 1.4360 to 1.4380
Quantity 250 mL
Beilstein 07,5
Merck Index 15,2738
Specific Gravity 0.95
Solubility Information Solubility in water: 301g/L (20°C). Other solubilities: miscible with alcohol,acetone and ether
Viscosity 0.8 mPa.s (20°C)
Formula Weight 84.12
Percent Purity 99+%
Grade Pure
Physical Form Liquid
Chemical Name or Material Cyclopentanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopentanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.