Learn More
Cyclopentyl bromide, 98%, Thermo Scientific Chemicals
£16.18 - £122.61
Chemical Identifiers
CAS | 137-43-9 |
---|---|
Molecular Formula | C5H9Br |
Molecular Weight (g/mol) | 149.03 |
MDL Number | MFCD00001359 |
InChI Key | BRTFVKHPEHKBQF-UHFFFAOYSA-N |
Synonym | cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane |
PubChem CID | 8728 |
IUPAC Name | bromocyclopentane |
SMILES | C1CCC(C1)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10294180
|
Thermo Scientific Acros
111600500 |
50 mL | Glass bottle |
£16.18
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10305890
|
Thermo Scientific
111602500 |
250 mL | Glass bottle |
£46.93
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10321564
|
Thermo Scientific Acros
111600010 |
1 L | Glass bottle |
£122.61
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
137-43-9 | |
149.03 | |
BRTFVKHPEHKBQF-UHFFFAOYSA-N | |
8728 | |
C1CCC(C1)Br |
C5H9Br | |
MFCD00001359 | |
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane | |
bromocyclopentane |
Specifications
137-43-9 | |
1.3900g/mL | |
35°C | |
97.5% min. (GC) | |
C5H9Br | |
MFCD00001359 | |
1.39 | |
Solubility in water: insoluble in water | |
C1CCC(C1)Br | |
149.03 | |
149.03 | |
Liquid |
Yellow to Brown | |
137°C to 139°C | |
Authentic | |
C5H9Br | |
1.4881 to 1.4901 | |
05, 19 | |
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane | |
BRTFVKHPEHKBQF-UHFFFAOYSA-N | |
bromocyclopentane | |
8728 | |
98% | |
Cyclopentyl bromide, 98% |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-294-6
missing translation for 'tsca' : TSCA
- Cyclopentyl bromide
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only