missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl isocyanide, 96%

Product Code. 11985701 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11985701

Brand: Thermo Scientific Acros 372090050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68498-54-4 | C6H9N | 95.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68498-54-4
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.14
InChI Key XTLSDEJNOQVCHL-UHFFFAOYSA-N
Synonym cyclopentyl isocyanide, cyclopentylisocyanide, isocyano-cyclopentan, isocyano-cyclopentane, cyclopentyl isonitrile, 3-isocyano-cyclopentane, acmc-1bgsw, tos-bb-0762
PubChem CID 2734784
IUPAC Name isocyanocyclopentane
SMILES [C-]#[N+]C1CCCC1

Specifications

Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 40°C (11.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4390 to 1.4410
Quantity 5 g
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble
Formula Weight 95.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Cyclopentyl isocyanide
Show More Show Less
compliance-icons
Product Identifier
  • Cyclopentyl isocyanide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.