missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized, Thermo Scientific Chemicals

Product Code. 10589213
Click to view available options
Quantity:
50 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5614-37-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD08276401
InChI Key SKTCDJAMAYNROS-UHFFFAOYSA-N
Synonym cyclopentyl methyl ether, cyclopentane, methoxy, cpme, cyclopentane,methoxy, unii-4067e5gbkb, cyclopentylmethylether, cylopentylmethylether, methoxycyclopentane, 2-methoxycyclopentyl, cylopentyl-methylether
PubChem CID 138539
IUPAC Name methoxycyclopentane
SMILES COC1CCCC1

Specifications

Melting Point -140.0°C
Density 0.8600g/mL
Boiling Point 106.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 99.9% min. (GC)
Packaging Glass bottle
Linear Formula C5H9OCH3
Refractive Index 1.4190 to 1.4220 (20°C, 589nm)
Quantity 50 mL
Residue after Evaporation 0.01% max.
Merck Index 15,2742
Specific Gravity 0.86
Stabilizer 50ppm (BHT) approximately
Formula Weight 100.16
Percent Purity 99.9+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Cyclopentyl methyl ether, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopentyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.