missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentyl methyl ether, Reagent Grade, contains 50 ppm BHT as inhibitor, ≥99.90%, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
5 L
Packaging:
Aluminum bottle
Glass bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | -140°C |
Density | 0.86g/mL at 25°C |
Boiling Point | 106°C (1013 hPa) |
Packaging | Aluminum bottle |
Quantity | 5 L |
UN Number | UN3271 |
Formula Weight | 100.16g/mol |
Percent Purity | ≥99.90% |
Chemical Name or Material | Cyclopentyl methyl ether |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'