missing translation for 'onlineSavingsMsg'
Learn More
Cyclopentyl thiocyanate, 95%, Thermo Scientific™ Product Code.: 15444927

Cyclopentyl thiocyanate, 95%, Thermo Scientific™

Product Code. 15444927
500 mg
Click to view available options
Quantity:
500 mg
2 g
This item is not returnable. View return policy

Product Code. 15444927

Brand: Thermo Scientific Alfa Aesar H32230.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5263-57-0
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.21
MDL Number MFCD11100558
InChI Key ZTZNTHBAUFZQGQ-UHFFFAOYSA-N
Synonym cyclopentanethiocyanate, cyclopentylsulfanyl formonitrile, cyclopentylrhodanid
PubChem CID 12376019
IUPAC Name cyclopentyl thiocyanate
SMILES N#CSC1CCCC1

Specifications

Boiling Point 84°C (8 mmHg)
Quantity 500 mg
Formula Weight 127.21
Percent Purity 95%
Chemical Name or Material Cyclopentyl thiocyanate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10160180 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclopentyl thiocyanate, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.