missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylacetylene, 95%, Thermo Scientific Chemicals

Product Code. 10411723
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-51-8
Molecular Formula C7H10
Molecular Weight (g/mol) 94.16
MDL Number MFCD00013744
InChI Key TXVJSWLZYQMWPC-UHFFFAOYSA-N
Synonym cyclopentylacetylene, cyclopentyl acetylene, cyclopentylethyne, cyclopentane, ethynyl, cyclopentane, ethynyl-7ci, 8ci, 9ci, ethynylcylopentane, ethynyl-cyclopentane, cyclopentyl-acetylen
PubChem CID 136725
IUPAC Name ethynylcyclopentane
SMILES C#CC1CCCC1

Specifications

Color Yellow
Boiling Point 107°C to 109°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH∽CC5H9
Refractive Index 1.4470 to 1.449
Quantity 1 g
Formula Weight 94.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Cyclopentylacetylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopentylacetylene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'