Learn More
Chemical Identifiers
CAS | 1003-03-8 |
---|---|
Molecular Formula | C5H11N |
Molecular Weight (g/mol) | 85.15 |
MDL Number | MFCD00001380 |
InChI Key | NISGSNTVMOOSJQ-UHFFFAOYSA-N |
Synonym | cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine |
PubChem CID | 2906 |
IUPAC Name | cyclopentanamine |
SMILES | NC1CCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10799181
|
Thermo Scientific Acros
111590050 |
5 g | Glass bottle |
£14.77
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10576251
|
Thermo Scientific Acros
111590250 |
25 g | Glass bottle |
£25.02
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10162641
|
Thermo Scientific Acros
111591000 |
100 g | Glass bottle |
£65.22
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10125203
|
Thermo Scientific Acros
111595000 |
500 g | Glass bottle |
£256.28
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1003-03-8 | |
85.15 | |
NISGSNTVMOOSJQ-UHFFFAOYSA-N | |
2906 | |
NC1CCCC1 |
C5H11N | |
MFCD00001380 | |
cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine | |
cyclopentanamine |
Specifications
1003-03-8 | |
106°C to 108°C | |
Authentic | |
C5H11N | |
1.4482 to 1.4502 | |
12,4 | |
cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine | |
NISGSNTVMOOSJQ-UHFFFAOYSA-N | |
cyclopentanamine | |
2906 | |
99+% | |
Cyclopentylamine, 99+% |
0.8600g/mL | |
17°C | |
99% min. (GC) | |
C5H9NH2 | |
MFCD00001380 | |
0.86 | |
Solubility in water: completely miscible with water | |
NC1CCCC1 | |
85.15 | |
85.15 | |
Liquid |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-697-3
missing translation for 'rtecsNumber' : GY8452000
missing translation for 'tsca' : TSCA
- Cyclopentylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Cyclopentanamine
RUO – Research Use Only