missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentylamine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Cyclopentylamine acts as an important raw material and intermediate used in organic synthesis and pharmaceuticals. Further, it serves as chemokine receptor 2 antagonists. It acts as a ligand in coordination chemistry and forms cis-dichlorobis(cyclopentylamine)platinum(II) complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopentylamine acts as an important raw material and intermediate used in organic synthesis and pharmaceuticals. Further, it serves as chemokine receptor 2 antagonists. It acts as a ligand in coordination chemistry and forms cis-dichlorobis(cyclopentylamine)platinum(II) complex.
Solubility
Miscible with water, acetone and benzene.
Notes
Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and carbon dioxide.
Cyclopentylamine acts as an important raw material and intermediate used in organic synthesis and pharmaceuticals. Further, it serves as chemokine receptor 2 antagonists. It acts as a ligand in coordination chemistry and forms cis-dichlorobis(cyclopentylamine)platinum(II) complex.
Solubility
Miscible with water, acetone and benzene.
Notes
Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and carbon dioxide.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -85°C |
Density | 0.862 |
pH | 12.5 |
Boiling Point | 106°C to 108°C |
Flash Point | 12°C (53°F) |
Odor | Amine-like |
Refractive Index | 1.4505 |
Quantity | 100 g |
UN Number | UN3286 |
Beilstein | 635706 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'