missing translation for 'onlineSavingsMsg'
Learn More
Cyclopentylboronic acid, 95%, Thermo Scientific™ Product Code.: 11925831

Cyclopentylboronic acid, 95%, Thermo Scientific™

Product Code. 11925831
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 11925831

Brand: Thermo Scientific Acros 359060010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 63076-51-7
Molecular Formula C5H11BO2
Molecular Weight (g/mol) 113.95
InChI Key VTTDFSNKIMAQTB-UHFFFAOYSA-N
Synonym cyclopentaneboronic acid, boronic acid, cyclopentyl, cylopentyl boronic acid, cyclopentylboronicacid, pubchem7792, acmc-209ncu, --paclitaxel;paclitaxel, boronic acid, b-cyclopentyl, cyclopentylboronic acid
PubChem CID 2734327
IUPAC Name cyclopentylboronic acid
SMILES B(C1CCCC1)(O)O

Specifications

Melting Point 134.0°C to 142.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 113.95
Percent Purity 95%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Cyclopentylboronic acid
compliance-icons
Product Identifier
  • Cyclopentylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclopentylboronic acid, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.