missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10555324
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33240-34-5, 60-29-7
Molecular Formula C5H9BrMg
Molecular Weight (g/mol) 173.34
MDL Number MFCD00209569
InChI Key PZOJUJCALDFFHC-UHFFFAOYSA-M
Synonym cyclopentylmagnesium bromide, bromocyclopentylmagnesium, magnesium, bromocyclopentyl, cyclopentyl magnesium bromide, bromo cyclopentyl magnesium, cyclopentylmagnesium bromide solution, cydopentylmagnesium bromide, cyclopentylmagnsium bromide, pzojujcaldffhc-uhfffaoysa-m, cyclopentylmagnesium bromide solution, 2.0 m in diethyl ether, grignard reagent
PubChem CID 118418
IUPAC Name magnesium;cyclopentane;bromide
SMILES Br[Mg]C1CCCC1

Specifications

Color Gray
Density 0.9960g/mL
Boiling Point 35°C
Flash Point -38°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.996
Formula Weight 173.33
Physical Form Solution
Chemical Name or Material Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H261-In contact with water releases flammable gases.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.