missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylmagnesium chloride, 1M in MeTHF, Thermo Scientific™

Product Code. 15498047
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32916-51-1
Molecular Formula C5H9ClMg
Molecular Weight (g/mol) 128.88
MDL Number MFCD00001357
InChI Key GROGEABZCKQTJW-UHFFFAOYSA-M
Synonym cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium
PubChem CID 3599845
SMILES Cl[Mg]C1CCCC1

Specifications

Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
IUPAC Name magnesium(2+) cyclopentanide chloride
Formula Weight 128.88
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material Cyclopentylmagnesium chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'