missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropanecarbonyl chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
Solubility
Soluble in water (reacts violently).
Notes
Moisture Sensitive. Store away from oxidizing agents, bases, active metals and water/moisture. Store under inert gas.
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
Solubility
Soluble in water (reacts violently).
Notes
Moisture Sensitive. Store away from oxidizing agents, bases, active metals and water/moisture. Store under inert gas.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.15 |
Boiling Point | 119°C to 120°C |
Flash Point | 23°C (73°F) |
Odor | Pungent |
Refractive Index | 1.452 |
Quantity | 25 g |
UN Number | UN2920 |
Beilstein | 471286 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water (reacts violently). |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'