missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanemethylamine, 97%, Thermo Scientific Chemicals

£22.90 - £254.00

Chemical Identifiers

CAS 2516-47-4
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.123
MDL Number MFCD00037147
InChI Key IGSKHXTUVXSOMB-UHFFFAOYSA-N
Synonym cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine
PubChem CID 75646
IUPAC Name cyclopropylmethanamine
SMILES C1CC1CN
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11491678
View Documents
Thermo Scientific Alfa Aesar
A13946.03
1 g
£22.90
1g
Please to purchase this item. Need a web account? Register with us today!
11401688
View Documents
Thermo Scientific Alfa Aesar
A13946.06
5 g
£63.30
5g
Please to purchase this item. Need a web account? Register with us today!
11411688
View Documents
Thermo Scientific Alfa Aesar
A13946.14
25 g
£254.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, air , carbon dioxide.
Specifications

Chemical Identifiers

2516-47-4
71.123
IGSKHXTUVXSOMB-UHFFFAOYSA-N
75646
C1CC1CN
C4H9N
MFCD00037147
cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine
cyclopropylmethanamine

Specifications

2516-47-4
85°C to 86°C
Amine-like
1.433
UN2733
Air Sensitive
Fully miscible in water.
C1CC1CN
71.123
71.12
Cyclopropanemethylamine
0.82
2°C (36°F)
C4H9N
MFCD00037147
773674
cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine
IGSKHXTUVXSOMB-UHFFFAOYSA-N
cyclopropylmethanamine
75646
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 219-737-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only