missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10496292
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 56601-42-4
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.15
MDL Number MFCD00040874
InChI Key JGFBQFKZKSSODQ-UHFFFAOYSA-N
Synonym cyclopropyl isothiocyanate, cyclopropane, isothiocyanato, isothiocyanato-cyclopropane, cyclopropylisothiocyanate, acmc-20albt, pubchem22785, cyclopropyl iso-thiocyanate, cyclopropane,isothiocyanato
PubChem CID 92463
IUPAC Name isothiocyanatocyclopropane
SMILES S=C=NC1CC1

Specifications

CAS Min % 97.0
Density 1.07g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula C3H5NCS
Quantity 1 g
Formula Weight 99.16
Percent Purity 97%
Chemical Name or Material Cyclopropyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'