missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylamine, 99%, Thermo Scientific Chemicals

Product Code. 10163250
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-30-0
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.09
InChI Key HTJDQJBWANPRPF-UHFFFAOYSA-N
Synonym cyclopropylamine, aminocyclopropane, cyclopropyl amine, unii-8pr8xth1x1, 8pr8xth1x1, cyclopropanamin, cyclopropyamine, cyclopropylamin, cylopropylamine, cycloproplyamine
PubChem CID 69828
ChEBI CHEBI:34660
IUPAC Name cyclopropanamine
SMILES C1CC1N

Specifications

Melting Point -50.0°C
Color Colorless
Density 0.8240g/mL
Boiling Point 49.0°C to 50.0°C
Flash Point −25°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C3H5NH2
Refractive Index 1.4190 to 1.4220
Quantity 10 mL
Beilstein 12,3
Specific Gravity 0.824
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 57.09
Percent Purity 99%
Physical Form Volatile Liquid
Chemical Name or Material Cyclopropylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'