missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropylboronic acid, tech. 85%, Thermo Scientific Chemicals
£55.70 - £195.00
Chemical Identifiers
CAS | 411235-57-9 |
---|---|
Molecular Formula | C3H7BO2 |
Molecular Weight (g/mol) | 85.90 |
MDL Number | MFCD04038750 |
InChI Key | WLVKDFJTYKELLQ-UHFFFAOYSA-N |
Synonym | cyclopropyl boronic acid, cyclopropylboranediol, cyclopropaneboronic acid, boronic acid, cyclopropyl, cyclopropyl-boronic acid, boronic acid, cyclopropyl-9ci, cyclopropylboronicacid, boronocyclopropane, cyclopropyboronic acid, cyclopropylboromc acid |
PubChem CID | 2760897 |
IUPAC Name | cyclopropylboronic acid |
SMILES | OB(O)C1CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11376719
|
Thermo Scientific Alfa Aesar
H26254.03 |
1 g |
£55.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386719
|
Thermo Scientific Alfa Aesar
H26254.06 |
5 g |
£195.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
411235-57-9 | |
85.90 | |
WLVKDFJTYKELLQ-UHFFFAOYSA-N | |
2760897 | |
OB(O)C1CC1 |
C3H7BO2 | |
MFCD04038750 | |
cyclopropyl boronic acid, cyclopropylboranediol, cyclopropaneboronic acid, boronic acid, cyclopropyl, cyclopropyl-boronic acid, boronic acid, cyclopropyl-9ci, cyclopropylboronicacid, boronocyclopropane, cyclopropyboronic acid, cyclopropylboromc acid | |
cyclopropylboronic acid |
Specifications
411235-57-9 | |
C3H7BO2 | |
cyclopropyl boronic acid, cyclopropylboranediol, cyclopropaneboronic acid, boronic acid, cyclopropyl, cyclopropyl-boronic acid, boronic acid, cyclopropyl-9ci, cyclopropylboronicacid, boronocyclopropane, cyclopropyboronic acid, cyclopropylboromc acid | |
OB(O)C1CC1 | |
85.90 | |
85.9 | |
Technical |
90°C to 95°C | |
MFCD04038750 | |
WLVKDFJTYKELLQ-UHFFFAOYSA-N | |
cyclopropylboronic acid | |
2760897 | |
85% | |
Cyclopropylboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only