Learn More
Cytiva CyDye™ Fluors Fluorophores
Tolerant of pH levels that range from 3-10 for use in a variety of applications at biologically relevant pHs
Brand: Cytiva 25-9004-72
Code : GE
Additional Details : Weight : 0.01600kg
Description
- Dyes are DMSO tolerant and photostable
- QC tested
Specifications
Red | |
NHS ester Cy3 | |
150 μg |
3 to 10 | |
Solid |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1