Learn More
Thermo Scientific Chemicals Cylindrical Al-24 Crucibles
£42.21 - £382.91
Specifications
Material Al-24
Quantity 1 Piece
Product Type Specs Cylindrical Al-24 Crucibles
View More Specs
Products 7
Product Code Brand Diameter (Metric) Outer Diameter (Metric) Inner Height (Metric) Capacity (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Diameter (Metric) Inner Height (Metric) Capacity (Metric) Price Quantity & Availability  
15483515
View Documents
Thermo Scientific Chemicals
032961.KF
16mm 30mm 20mm 6 mL
£42.21
Each
In Stock
to see stock.
Add to basket
15493515
View Documents
Thermo Scientific Chemicals
032962.KF
26mm 40mm 30mm 20 mL
£102.51
Each
In Stock
to see stock.
Add to basket
15403525
View Documents
Thermo Scientific Chemicals
032963.KF
30mm 50mm 35mm 40 mL
£100.50
Each
In Stock
to see stock.
Add to basket
15413525
View Documents
Thermo Scientific Chemicals
032964.KF
36mm 60mm 40mm 60 mL
£215.00
Each
In Stock
to see stock.
Add to basket
15423525
View Documents
Thermo Scientific Chemicals
032965.KF
50mm 75mm 75mm 110 mL
£181.91
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
15433525
View Documents
Thermo Scientific Chemicals
032966.KF
55mm 100mm 65mm 225 mL
£382.91
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
15443525
View Documents
Thermo Scientific Chemicals
032967.KF
73mm 150mm 85mm 600 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Al-24
1 Piece
Cylindrical Al-24 Crucibles