missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Cylindrical Concave Mirrors
Cylindrical concave mirrors are stainless-steel with polished composition.
Brand: Nickel Electro™ XFK-530-D
This item is not returnable.
View return policy
Description
- Stainless Steel Concave Mirrors
Order Info
Size: 165 mm diameter. Sold in packs of 4.
Specifications
4 Pk. | |
165 mm | |
Concave Mirror |
Stainless Steel | |
6.5 in. | |
Concave |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nickel Electro™ Cylindrical Concave Mirrors > Diameter: 165 mm, Quantity: 4 Pk.
Spot an opportunity for improvement?Share a Content Correction