Learn More
Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels, with PTFE Plug
Useful for transferring fluids and handling air-sensitive reagents in a sealed, inert-gas environment. Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
£108.12 - £614.78
Specifications
Material | Borosilicate Glass |
---|---|
Graduated | Yes |
Product Type | Dropping Funnel |
Product Code | Brand | Capacity (Metric) | Joint Number | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Joint Number | Description | Price | Quantity & Availability | |||
10589041
|
Lenz Laborglasinstrumente™
04377014 |
1000 mL | 29/32 NS | Pressure Equalizing Tube |
£614.78
Pack of 2 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
10047810
|
Lenz Laborglasinstrumente™
04177014 |
1000 mL | 29/32 NS | Without Pressure Equalizing Tube |
£220.00
Pack of 2 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
10635041
|
Lenz Laborglasinstrumente™
04373711 |
100 mL | 14/23 NS | Pressure Equalizing Tube |
£376.30
Pack of 2 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10343001
|
Lenz Laborglasinstrumente™
04173714 |
100 mL | 29/32 NS | Without Pressure Equalizing Tube |
£129.00
Pack of 2 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
10500012
|
Lenz Laborglasinstrumente™
04174914 |
250 mL | 29/32 NS | Without Pressure Equalizing Tube |
£143.00
Pack of 2 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
10519241
|
Lenz Laborglasinstrumente™
04172811 |
25 mL | 14/23 NS | Without Pressure Equalizing Tube |
£116.00
Pack of 2 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
10740651
|
Lenz Laborglasinstrumente™
04374914 |
250 mL | 29/32 NS | Pressure Equalizing Tube |
£464.28
Pack of 2 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||
10149650
|
Lenz Laborglasinstrumente™
04175814 |
500 mL | 29/32 NS | Without Pressure Equalizing Tube |
£170.00
Pack of 2 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
10707711
|
Lenz Laborglasinstrumente™
04372211 |
25 mL | 14/23 NS | Pressure Equalizing Tube |
£146.00
Pack of 2 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
12586229
|
Lenz Laborglasinstrumente™
04372811 |
50 mL | 14/23 NS | Pressure Equalizing Tube |
£316.94
Pack of 2 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
10590002
|
Lenz Laborglasinstrumente™
04172814 |
50 mL | 29/32 NS | Without Pressure Equalizing Tube |
£116.00
Pack of 2 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
10792132
|
Lenz Laborglasinstrumente™
04375814 |
500 mL | 29/32 NS | Pressure Equalizing Tube |
£224.98
Pack of 2 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
12556229
|
Lenz Laborglasinstrumente™
04372814 |
50 mL | 29/32 NS | Pressure Equalizing Tube |
£108.12
Pack of 2 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
Description
- With scale (approx. volume/mL)
- With screwthread retaining device
- Cylindrical shape with NS socket and cone
- Corrosion-free and resistant to thermal shock
Specifications
Borosilicate Glass | |
Yes | |
Dropping Funnel |